3-Bromo-4-butoxybenzoic acid

Supplier: Matrix Scientific

024092-500MG
101817-396EA 306.15 USD
101817-396
3-Bromo-4-butoxybenzoic acid
3-Bromo-4-butoxybenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₃BrO₃
MDL Number: MFCD06662499
CAS Number: 450416-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR