2-(4-Isopropylphenoxy)propanoic acid

Supplier: Matrix Scientific

024091-500MG
101817-394EA 306.15 USD
101817-394
2-(4-Isopropylphenoxy)propanoic acid
2-(4-Isopropylphenoxy)propanoic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₃
MDL Number: MFCD03422222
CAS Number: 237412-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR