(2-Nitrophenoxy)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(2-Nitrophenoxy)acetic acid

024083-500MG
101817-374EA 15.38 USD
101817-374
(2-Nitrophenoxy)acetic acid
(2-Nitrophenoxy)acetic acid

Hazard: Irritant

Formula: (C₆H₄NO₂)OCH₂COOH
MW: 197.15 g/mol
MDL Number: MFCD00016993
CAS Number: 1878-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR