2-([1,1'-Biphenyl]-4-yloxy)acetic acid

Supplier: Matrix Scientific

024076-500MG
101817-360EA 284.62 USD
101817-360
2-([1,1'-Biphenyl]-4-yloxy)acetic acid
2-([1,1'-Biphenyl]-4-yloxy)acetic acid

Hazard: Irritant

Formula: C₁₄H₁₂O₃
MDL Number: MFCD00179413
CAS Number: 13333-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR