2-(2-Methylphenoxy)propanoic acid

Supplier: Matrix Scientific

Synonyms: 2-(o-Tolyloxy)propanoic acid

024068-500MG
101817-344EA 306.15 USD
101817-344
2-(2-Methylphenoxy)propanoic acid
2-(2-Methylphenoxy)propanoic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
MDL Number: MFCD01122029
CAS Number: 7345-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR