(3,4-Xylyloxy)acetic acid

Supplier: Matrix Scientific

024066-500MG
101817-340EA 15.38 USD
101817-340
(3,4-Xylyloxy)acetic acid
(3,4-Xylyloxy)acetic acid

Hazard: Irritant

Formula: (C₆H₃(CH₃)₂)OCH₂COOH
MW: 180.2 g/mol
MDL Number: MFCD00017595
CAS Number: 13335-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR