2-(4-Methylphenoxy)propanoic acid

Supplier: Matrix Scientific

024065-500MG
101817-338EA 129.23 USD
101817-338
2-(4-Methylphenoxy)propanoic acid
2-(4-Methylphenoxy)propanoic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MDL Number: MFCD02684121
CAS Number: 22504-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR