2-(4-Bromo-2-chlorophenoxy)acetic acid

Supplier: Matrix Scientific

024062-500MG
101817-332EA 306.15 USD
101817-332
2-(4-Bromo-2-chlorophenoxy)acetic acid
2-(4-Bromo-2-chlorophenoxy)acetic acid

Hazard: Irritant

Formula: C₈H₆BrClO₃
MDL Number: MFCD03422201
CAS Number: 77228-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR