2-(2-Bromo-4-ethylphenoxy)acetic acid

Supplier: Matrix Scientific

024060-500MG
101817-328EA 495.38 USD
101817-328
2-(2-Bromo-4-ethylphenoxy)acetic acid
2-(2-Bromo-4-ethylphenoxy)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₁BrO₃
MDL Number: MFCD03488754
CAS Number: 90841-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR