2-(4-Ethylphenoxy)acetic acid

Supplier: Matrix Scientific

024058-500MG
101817-320EA 15.38 USD
101817-320
2-(4-Ethylphenoxy)acetic acid
2-(4-Ethylphenoxy)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00014366
CAS Number: 24431-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR