2-(4-Bromo-2-methylphenoxy)acetic acid

Supplier: Matrix Scientific

024056-500MG
101817-318EA 306.15 USD
101817-318
2-(4-Bromo-2-methylphenoxy)acetic acid
2-(4-Bromo-2-methylphenoxy)acetic acid

Hazard: Irritant

Formula: C₉H₉BrO₃
MDL Number: MFCD03129271
CAS Number: 6956-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR