2-(4-Bromo-3-methylphenoxy)acetic acid

Supplier: Matrix Scientific

024054-500MG
101817-314EA 196.92 USD
101817-314
2-(4-Bromo-3-methylphenoxy)acetic acid
2-(4-Bromo-3-methylphenoxy)acetic acid

Hazard: Irritant

Formula: C₉H₉BrO₃
Melting Pt: 178 °C
MDL Number: MFCD03129268
CAS Number: 90971-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR