2-(2-Isopropyl-5-methylphenoxy)acetic acid

Supplier: Matrix Scientific

Synonyms: (2-Isopropyl-5-methylphenoxy)acetic acid

024053-500MG
101817-312EA 29.23 USD
101817-312
2-(2-Isopropyl-5-methylphenoxy)acetic acid
2-(2-Isopropyl-5-methylphenoxy)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
MDL Number: MFCD00021757
CAS Number: 5333-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR