3-(2,4-Dichlorophenoxy)propionic acid

Supplier: Matrix Scientific

Synonyms: 2-(2,4-Dichlorophenoxy)propionic acid

024043-500MG
101817-288EA 15.38 USD
101817-288
3-(2,4-Dichlorophenoxy)propionic acid
3-(2,4-Dichlorophenoxy)propionic acid

Hazard: Irritant

Formula: C₉H₈Cl₂O₃
MDL Number: MFCD00002645
CAS Number: 120-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR