2-[(1-Bromo-2-naphthyl)oxy]acetic acid

Supplier: Matrix Scientific

024042-500MG
101817-286EA 306.15 USD
101817-286
2-[(1-Bromo-2-naphthyl)oxy]acetic acid
2-[(1-Bromo-2-naphthyl)oxy]acetic acid

Hazard: Irritant

Formula: C₁₂H₉BrO₃
MDL Number: MFCD00453999
CAS Number: 41791-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR