2-[4-(sec-Butyl)phenoxy]acetic acid

Supplier: Matrix Scientific

024041-500MG
101817-284EA 306.15 USD
101817-284
2-[4-(sec-Butyl)phenoxy]acetic acid
2-[4-(sec-Butyl)phenoxy]acetic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₃
MDL Number: MFCD02631950
CAS Number: 4917-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR