MATR024038

024038-500MG
101817-278EA 18.31 USD
101817-278
MATR024038
CAS: 1643-15-8
MDL Number: MFCD00014358
Molecular Formula: C9H10O3
Molecular Weight: 166,18
Hazard: Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR