2-[2-(sec-Butyl)phenoxy]acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(2-(sec-Butyl)phenoxy)acetic acid

024030-500MG
101817-262EA 306.15 USD
101817-262
2-[2-(sec-Butyl)phenoxy]acetic acid
2-[2-(sec-Butyl)phenoxy]acetic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
MDL Number: MFCD00268165
CAS Number: 76343-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR