2-Phenylbenzoic acid

Supplier: Matrix Scientific

Synonyms: [1,1'-Biphenyl]-2-carboxylic acidBiphenyl-2-carboxylic acid2-Biphenylcarboxylic acid

027210-1G
101823-736EA 11.5 USD
101823-736
2-Phenylbenzoic acid
2-Phenylbenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
MDL Number: MFCD00002463
CAS Number: 947-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR