1H-Pyrazole-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Pyrazolecarboxylic acid

027208-1G 027208-25G
101823-732EA 95.38 USD
101823-732 101823-728
1H-Pyrazole-4-carboxylic acid ≥97%
1H-Pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: 282 °C
MDL Number: MFCD00011558
CAS Number: 37718-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR