(1,3-Benzoxazol-2-ylthio)acetic acid

Supplier: Matrix Scientific

027195-500MG
101823-694EA 290.77 USD
101823-694
(1,3-Benzoxazol-2-ylthio)acetic acid
(1,3-Benzoxazol-2-ylthio)acetic acid

Hazard: Irritant

Formula: C₉H₇NO₃S
MDL Number: MFCD00463910
CAS Number: 58089-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR