4-Methyl-1H-imidazole-5-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 5-Methyl-4-imidazolecarboxylic Acid

027183-500MG
101823-676EA 138.46 USD
101823-676
4-Methyl-1H-imidazole-5-carboxylic acid
4-Methyl-1H-imidazole-5-carboxylic acid

Hazard: Irritant

Formula: C₅H₆N₂O₂
MW: 126.12 g/mol
MDL Number: MFCD00233368
CAS Number: 1457-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR