4-Isoquinolinecarboxylic acid

Supplier: Matrix Scientific

Synonyms: Isoquinoline-4-carboxylic acid

027181-500MG
101823-672EA 289.8 USD
101823-672
4-Isoquinolinecarboxylic acid
4-Isoquinolinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₂
MW: 173.1681 g/mol
Melting Pt: 264…266 °C
MDL Number: MFCD00094344
CAS Number: 7159-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR