2-(4-Ethylphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027176-500MG
101823-660EA 146.15 USD
101823-660
2-(4-Ethylphenyl)quinoline-4-carboxylic acid
2-(4-Ethylphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₅NO₂
MDL Number: MFCD00781757
CAS Number: 301320-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR