2-(4-Bromophenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027172-500MG
101823-652EA 138.46 USD
101823-652
2-(4-Bromophenyl)quinoline-4-carboxylic acid
2-(4-Bromophenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₀BrNO₂
MDL Number: MFCD00160560
CAS Number: 103914-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR