5-Oxothiomorpholine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Oxo-thiomorpholine-3-carboxylic acid

027167-500MG
101823-640EA 307.69 USD
101823-640
5-Oxothiomorpholine-3-carboxylic acid ≥95%
5-Oxothiomorpholine-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₇NO₃S
MW: 161.18 g/mol
Melting Pt: 182…183 °C
MDL Number: MFCD00671711
CAS Number: 14226-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR