2-Methyl-1,3-benzothiazole-6-carboxylic acid

Supplier: Matrix Scientific

027164-500MG
101823-634EA 387.69 USD
101823-634
2-Methyl-1,3-benzothiazole-6-carboxylic acid
2-Methyl-1,3-benzothiazole-6-carboxylic acid

Hazard: Irritant

Formula: C₉H₇NO₂S
MDL Number: MFCD07394023
CAS Number: 6941-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR