1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

027163-500MG
101823-632EA 50.77 USD
101823-632
1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid
1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
MDL Number: MFCD00159648
CAS Number: 1125-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR