2-Morpholin-4-ylpropanoic acid

Supplier: Matrix Scientific

027161-500MG
101823-630EA 290.77 USD
101823-630
2-Morpholin-4-ylpropanoic acid
2-Morpholin-4-ylpropanoic acid

Hazard: Irritant

Formula: C₇H₁₃NO₃
MDL Number: MFCD03821872
CAS Number: 161907-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR