2-Pyrrolidin-1-ylpropanoic acid

Supplier: Matrix Scientific

Synonyms: 2-(1-Pyrrolidyl)propanoic acid

027158-500MG
101823-624EA 350.77 USD
101823-624
2-Pyrrolidin-1-ylpropanoic acid
2-Pyrrolidin-1-ylpropanoic acid

Hazard: Irritant

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
MDL Number: MFCD05237232
CAS Number: 123912-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR