3-(Dimethylamino)propanoic acid ≥97%

Supplier: Matrix Scientific

027155-5G
101823-616EA 138.46 USD
101823-616
3-(Dimethylamino)propanoic acid ≥97%
3-(Dimethylamino)propanoic acid

Hazard: Irritant

Formula: C₅H₁₁NO₂
Boiling Pt: 140…142 °C (0.15 Torr)
Melting Pt: 143…144 °C
MDL Number: MFCD00482234
CAS Number: 6300-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR