(3-Oxopiperazin-2-yl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(3-Oxopiperazin-2-yl)acetic acid

027150-500MG
101823-606EA 290.77 USD
101823-606
(3-Oxopiperazin-2-yl)acetic acid
(3-Oxopiperazin-2-yl)acetic acid

Hazard: Irritant

Formula: C₆H₁₀N₂O₃
MW: 158.16 g/mol
MDL Number: MFCD04226363
CAS Number: 405214-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR