5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

027148-500MG
101823-602EA 290.77 USD
101823-602
5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid
5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉N₃O₂
MDL Number: MFCD00220607
CAS Number: 51649-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR