3-(Chloromethyl)-4-methoxybenzaldehyde

Supplier: Matrix Scientific

027147-500MG
101823-600EA 146.15 USD
101823-600
3-(Chloromethyl)-4-methoxybenzaldehyde
3-(Chloromethyl)-4-methoxybenzaldehyde

Hazard: Irritant

Formula: C₉H₉ClO₂
MDL Number: MFCD00995681
CAS Number: 52577-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR