1-Isopropyl-1H-pyrazole-4-carbaldehyde

Supplier: Matrix Scientific

027145-500MG
101823-598EA 396.92 USD
101823-598
1-Isopropyl-1H-pyrazole-4-carbaldehyde
1-Isopropyl-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₇H₁₀N₂O
MDL Number: MFCD06260724
CAS Number: 313735-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR