4-Formylbenzenesulfonamide

Supplier: Matrix Scientific

027134-500MG
101823-574EA 223.08 USD
101823-574
4-Formylbenzenesulfonamide
4-Formylbenzenesulfonamide

Hazard: Irritant

Formula: C₇H₇NO₃S
MDL Number: MFCD03196508
CAS Number: 3240-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR