5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde

Supplier: Matrix Scientific

Synonyms: 5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde

027130-500MG
101823-564EA 174.8 USD
101823-564
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₉H₁₁ClN₂O
MW: 220.65 g/mol
MDL Number: MFCD00100755
CAS Number: 947-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR