2-Piperidin-2-yl-1H-benzimidazole dihydrochloride

Supplier: Matrix Scientific

Synonyms: 2-(Piperidin-2-yl)-1H-benzo[d]imidazole

027111-500MG
101823-518EA 461.54 USD
101823-518
2-Piperidin-2-yl-1H-benzimidazole dihydrochloride
2-Piperidin-2-yl-1H-benzimidazole dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂N₃
MW: 274.2 g/mol
MDL Number: MFCD08461220
CAS Number: 51785-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR