N-Methyl-N-[(5-methyl-1,3,4-oxadiazol-2-yl)methyl]amine hydrochloride

Supplier: Matrix Scientific

027109-500MG
101823-514EA 461.54 USD
101823-514
N-Methyl-N-[(5-methyl-1,3,4-oxadiazol-2-yl)methyl]amine hydrochloride
N-Methyl-N-[(5-methyl-1,3,4-oxadiazol-2-yl)methyl]amine hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClN₃O
MDL Number: MFCD11845787
CAS Number: 887405-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR