Tetrahydrothien-3-ylamine

Supplier: Matrix Scientific

027088-500MG
101823-466EA 560 USD
101823-466
Tetrahydrothien-3-ylamine
Tetrahydrothien-3-ylamine

Hazard: Irritant

Formula: C₄H₉NS
MDL Number: MFCD00120988
CAS Number: 101993-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR