3-(Morpholinosulfonyl)aniline

Supplier: Matrix Scientific

027084-500MG
101823-458EA 193.85 USD
101823-458
3-(Morpholinosulfonyl)aniline
3-(Morpholinosulphonyl)aniline

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₃S
MDL Number: MFCD02090909
CAS Number: 22184-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR