4-Amino-N-(3,5-dichlorophenyl)benzenesulfonamide

Supplier: Matrix Scientific

027083-500MG
101823-456EA 193.85 USD
101823-456
4-Amino-N-(3,5-dichlorophenyl)benzenesulfonamide
4-Amino-N-(3,5-dichlorophenyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₀Cl₂N₂O₂S
MDL Number: MFCD00276375
CAS Number: 5407-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR