4-Amino-N-ethylbenzenesulfonamide

Supplier: Matrix Scientific

027079-500MG
101823-448EA 193.85 USD
101823-448
4-Amino-N-ethylbenzenesulfonamide
4-Amino-N-ethylbenzenesulphonamide

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD02090910
CAS Number: 1709-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR