2-(1-Methylpiperidin-2-yl)ethanamine
Supplier: Matrix Scientific
027075-500MG
101823-440EA
387.69
USD
101823-440
2-(1-Methylpiperidin-2-yl)ethanamine
2-(1-Methylpiperidin-2-yl)ethanamine
Hazard: Irritant
Formula:
C₈H₁₈N₂ |
MDL Number:
MFCD06408778 CAS Number: 18128-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...