(1-Methylpyrrolidin-2-yl)methylamine

Supplier: Matrix Scientific

027074-500MG
101823-438EA 243.08 USD
101823-438
(1-Methylpyrrolidin-2-yl)methylamine
(1-Methylpyrrolidin-2-yl)methylamine

Hazard: Irritant

Formula: C₆H₁₄N₂
MDL Number: MFCD09971798
CAS Number: 26171-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR