Thiomorpholine-3-carboxamide

Supplier: Matrix Scientific

027072-500MG
101823-434EA 387.69 USD
101823-434
Thiomorpholine-3-carboxamide
Thiomorpholine-3-carboxamide

Hazard: Irritant

Formula: C₅H₁₀N₂OS
MDL Number: MFCD02666349
CAS Number: 103742-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR