4-(1-Adamantyl)aniline

Supplier: Matrix Scientific

Synonyms: 4-(Adamantan-1-yl)aniline

027068-500MG
101823-430EA 387.69 USD
101823-430
4-(1-Adamantyl)aniline
4-(1-Adamantyl)aniline

Hazard: Irritant

Formula: C₁₆H₂₁N
MW: 227.35 g/mol
MDL Number: MFCD00193829
CAS Number: 1459-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR