N-1-Adamantyl-4-aminobenzenesulfonamide

Supplier: Matrix Scientific

027066-500MG
101823-426EA 193.85 USD
101823-426
N-1-Adamantyl-4-aminobenzenesulfonamide
N-1-Adamantyl-4-aminobenzenesulfonamide

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₂S
MDL Number: MFCD00276897
CAS Number: 24224-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR