1-Imidazo[1,2-a]pyridin-2-ylmethanamine dihydrochloride

Supplier: Matrix Scientific
Warning

027044-500MG
101823-382EA 387.69 USD
101823-382
1-Imidazo[1,2-a]pyridin-2-ylmethanamine dihydrochloride
1-Imidazo[1,2-a]pyridin-2-ylmethanamine dihydrochloride

Hazard: Irritant

Formula: C₈H₉N₃·2ClH
MW: 220.099 g/mol
MDL Number: MFCD04448842
CAS Number: 452967-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR