2-(5-Fluoro-1H-benzimidazol-2-yl)ethanamine dihydrochloride

Supplier: Matrix Scientific

027043-500MG
101823-380EA 387.69 USD
101823-380
2-(5-Fluoro-1H-benzimidazol-2-yl)ethanamine dihydrochloride
2-(5-Fluoro-1H-benzimidazol-2-yl)ethanamine dihydrochloride

Hazard: Irritant

Formula: C₉H₁₂Cl₂FN₃
MDL Number: MFCD09029275
CAS Number: 887405-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR